LIF372157569 Life Chemicals Inc.

N-(tert-butyl)-2-Chloroacetamide

  • MDL number MFCD00032565
  • Cas Number : 15678-99-6
  • Molecular Weight : 149.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart