LIF372157577 Life Chemicals Inc.

5-(2,5-dimethoxyphenyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11986944
  • Cas Number : 1105193-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart