LIF372157578 Life Chemicals Inc.

5-(m-tolyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD00469195
  • Cas Number : 109060-64-2
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart