LIF372157579 Life Chemicals Inc.

5-(1-benzofuran-2-yl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09940004
  • Cas Number : 933244-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart