LIF372157583 Life Chemicals Inc.

5-(5-chloro-2-thienyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11986949
  • Cas Number : 1105193-79-0
  • Molecular Weight : 201.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart