LIF372157584 Life Chemicals Inc.

5-[4-(methylthio)phenyl]-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11986950
  • Cas Number : 1105193-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart