LIF372157587 Life Chemicals Inc.

5-[3-(methylsulfonyl)phenyl]-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09804361
  • Cas Number : 1016764-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart