LIF372157588 Life Chemicals Inc.

5-[2-(methylsulfonyl)phenyl]-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11986952
  • Cas Number : 1105193-94-9
  • Molecular Weight : 239.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart