LIF372157589 Life Chemicals Inc.

4-(5-amino-1,3,4-oxadiazol-2-yl)-N,N-dimethylbenzenesulfonamide

  • MDL number MFCD11986953
  • Cas Number : 1105193-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart