LIF372157593 Life Chemicals Inc.

5-(2,4-dimethylbenzyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11986954
  • Cas Number : 1105194-04-4
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart