LIF372157595 Life Chemicals Inc.

5-(1,3-benzodioxol-5-ylmethyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11986955
  • Cas Number : 14731-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart