LIF372157601 Life Chemicals Inc.

7-chloro-4-methoxy-N-(pyridin-3-ylmethyl)benzo[d]thiazol-2-amine

  • MDL number MFCD11986960
  • Cas Number : 1105194-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart