LIF372157606 Life Chemicals Inc.

6-methyl-N-(tetrahydrofuran-2-ylmethyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD11986965
  • Cas Number : 1105194-49-7
  • Molecular Weight : 248.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart