LIF372157616 Life Chemicals Inc.

3-(1,3-benzothiazol-2-yl)-6-ethyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridin-2-amine

  • MDL number MFCD11986972
  • Cas Number : 1105194-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart