LIF372157622 Life Chemicals Inc.

methyl 2-amino-6-isopropyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate

  • MDL number MFCD01856353
  • Cas Number : 374102-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart