LIF372157623 Life Chemicals Inc.

N,N-dimethyl-N'-(6-methyl-1,3-benzothiazol-2-yl)ethane-1,2-diamine

  • MDL number MFCD11986976
  • Cas Number : 1105194-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart