LIF372157627 Life Chemicals Inc.

N'-(6-chloro-1,3-benzothiazol-2-yl)-N,N-diethylethane-1,2-diamine

  • MDL number MFCD11986980
  • Cas Number : 1105195-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart