LIF372157629 Life Chemicals Inc.

N'-(6-bromo-1,3-benzothiazol-2-yl)-N,N-dimethylethane-1,2-diamine

  • MDL number MFCD11986982
  • Cas Number : 1105195-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart