LIF372157638 Life Chemicals Inc.

N,N-diethyl-N'-(4-fluoro-1,3-benzothiazol-2-yl)ethane-1,2-diamine

  • MDL number MFCD11986989
  • Cas Number : 1105188-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart