LIF372157659 Life Chemicals Inc.

6-methoxy-N-(3-morpholinopropyl)benzo[d]thiazol-2-amine

  • MDL number MFCD11987009
  • Cas Number : 1105188-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart