LIF372157666 Life Chemicals Inc.

N'-(7-chloro-4-methoxy-1,3-benzothiazol-2-yl)-N,N-dimethylethane-1,2-diamine

  • MDL number MFCD11987016
  • Cas Number : 1105188-90-6
  • Molecular Weight : 285.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart