LIF372157667 Life Chemicals Inc.

N'-(7-chloro-4-methoxy-1,3-benzothiazol-2-yl)-N,N-dimethylpropane-1,3-diamine

  • MDL number MFCD11987017
  • Cas Number : 1105188-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart