LIF372157680 Life Chemicals Inc.

4,6-difluoro-N-(2-morpholin-4-ylethyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD11987029
  • Cas Number : 1105189-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart