LIF372157685 Life Chemicals Inc.

2-chloro-4-(methylsulfonyl)-1,3-benzothiazole

  • MDL number MFCD11987034
  • Cas Number : 1105189-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart