LIF372157695 Life Chemicals Inc.

4-(phenylthio)butanoic acid

  • MDL number MFCD02082831
  • Cas Number : 17742-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart