LIF372157697 Life Chemicals Inc.

4-(benzylsulfonyl)butanoic acid

  • MDL number MFCD11987038
  • Cas Number : 100059-54-9
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart