LIF372157702 Life Chemicals Inc.

2-(4-phenylpiperazin-1-yl)benzo[d]thiazol-6-ol

  • MDL number MFCD11987040
  • Cas Number : 1105189-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart