LIF372157706 Life Chemicals Inc.

4,4'-bi-1,3-thiazole-2,2'-diamine

  • MDL number MFCD01740587
  • Cas Number : 58139-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart