LIF372157710 Life Chemicals Inc.

4-(5-cyclopropyl-1,3,4-oxadiazol-2-yl)piperidine

  • MDL number MFCD11610657
  • Cas Number : 1082828-62-3
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart