LIF372157722 Life Chemicals Inc.

2-(chloromethyl)-2,3-dihydro-1,4-benzodioxine

  • MDL number MFCD00014666
  • Cas Number : 2164-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart