LIF372157729 Life Chemicals Inc.

4-(pyridin-2-yloxy)benzenesulfonyl chloride

  • MDL number MFCD09755965
  • Cas Number : 192329-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart