LIF372157732 Life Chemicals Inc.

ethyl (acetyloxy)(diethoxyphosphoryl)acetate

  • MDL number MFCD04039534
  • Cas Number : 162246-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart