LIF372157739 Life Chemicals Inc.

3-[3-(propan-2-yl)-1,2,4-oxadiazol-5-yl]propan-1-amine

  • MDL number MFCD09041879
  • Cas Number : 926230-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart