LIF372157740 Life Chemicals Inc.

{1-[(4-methylpiperazin-1-yl)methyl]propyl}amine

  • MDL number MFCD06446862
  • Cas Number : 84630-63-7
  • Molecular Weight : 171.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart