LIF372157741 Life Chemicals Inc.

2-fluoronaphthalene

  • MDL number MFCD00082618
  • Cas Number : 323-09-1
  • Molecular Weight : 146.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart