LIF372157743 Life Chemicals Inc.

N~1~,N~1~-dimethyl-1-(1-methyl-1H-indol-3-yl)ethane-1,2-diamine

  • MDL number MFCD01211520
  • Cas Number : 1225553-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart