LIF372157745 Life Chemicals Inc.

[2-(1-methyl-1H-pyrrol-2-yl)-2-pyrrolidin-1-ylethyl]amine

  • MDL number MFCD21092025
  • Cas Number : 1333960-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart