LIF372157761 Life Chemicals Inc.

2-(4-phenylpiperazin-1-yl)ethan-1-amine

  • MDL number MFCD03030396
  • Cas Number : 21091-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart