LIF372157780 Life Chemicals Inc.

(1-methyl-1H-pyrrol-2-yl)(morpholin-4-yl)acetonitrile

  • MDL number MFCD11987060
  • Cas Number : 80886-11-9
  • Molecular Weight : 205.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart