LIF372157805 Life Chemicals Inc.

3-(4-bromophenyl)pentanedioic acid

  • MDL number MFCD09032971
  • Cas Number : 1141-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart