LIF372157829 Life Chemicals Inc.

1-benzoyl-1,2,3,4-tetrahydroquinolin-6-amine

  • MDL number MFCD09261804
  • Cas Number : 857759-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart