LIF372157834 Life Chemicals Inc.

5-amino-1-methyl-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD09702413
  • Cas Number : 20870-91-1
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart