LIF372157842 Life Chemicals Inc.

8-amino-3,3-dimethyl-5-propyl-2,3-dihydro-1,5-benzoxazepin-4(5H)-one

  • MDL number MFCD11987126
  • Cas Number : 1171418-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart