LIF372157851 Life Chemicals Inc.

7-amino-4-methyl-3,4-dihydro-1,4-benzoxazepin-5(2H)-one

  • MDL number MFCD11987135
  • Cas Number : 129018-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart