LIF372157853 Life Chemicals Inc.

2-aminodibenzo[b,f][1,4]oxazepin-11(10H)-one

  • MDL number MFCD00660246
  • Cas Number : 23474-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart