LIF372157859 Life Chemicals Inc.

2-phenylmorpholine

  • MDL number MFCD01713554
  • Cas Number : 23972-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart