LIF372157874 Life Chemicals Inc.

2-(2-thienylcarbonyl)-1,2,3,4-tetrahydroisoquinolin-7-amine

  • MDL number MFCD10016679
  • Cas Number : 1017675-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart