LIF372157876 Life Chemicals Inc.

2-benzoyl-1,2,3,4-tetrahydroisoquinolin-7-amine

  • MDL number MFCD10016680
  • Cas Number : 855644-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart