LIF372157889 Life Chemicals Inc.

ethyl [(5-nitro-1,3-thiazol-2-yl)amino](oxo)acetate

  • MDL number MFCD00186394
  • Cas Number : 89792-36-9
  • Molecular Weight : 245.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart