LIF372157891 Life Chemicals Inc.

2-(thiophen-3-yl)ethan-1-amine

  • MDL number MFCD00176329
  • Cas Number : 59311-67-0
  • Molecular Weight : 127.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-thienyl)ethanamine
Inchi Code 1S/C6H9NS/c7-3-1-6-2-4-8-5-6/h2,4-5H,1,3,7H2
Inchi Key OANLWIQYRRVBPY-UHFFFAOYSA-N

Description

Purity 95%
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart